EMAIL THIS PAGE TO A FRIEND
336297 Sigma-Aldrich

Tributylantimony

technical grade, ≥90%

Purchase

Properties

grade   technical grade
assay   ≥90%
SMILES string   CCCC[Sb](CCCC)CCCC

Description

Packaging

25g units packaged under nitrogen in Sure/Seal bottles

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3394 4.3(4.2) / PGI
WGK Germany 
2
Flash Point(F) 
120.2 °F
Flash Point(C) 
49 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?