• USA Home
  • 336475 - 2,3,4-Trifluorobenzoyl chloride

EMAIL THIS PAGE TO A FRIEND
336475 Sigma-Aldrich

2,3,4-Trifluorobenzoyl chloride

98%

Purchase

Properties

assay   98%
refractive index   n20/D 1.497 (lit.)
bp   143-144 °C/130 mmHg (lit.)
density   1.522 g/mL at 25 °C (lit.)
SMILES string   Fc1ccc(C(Cl)=O)c(F)c1F
InChI   1S/C7H2ClF3O/c8-7(12)3-1-2-4(9)6(11)5(3)10/h1-2H
InChI key   NXRQXCFBZGIRGN-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3265 8 / PGII
WGK Germany 
3
Flash Point(F) 
199.4 °F
Flash Point(C) 
93 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?