• USA Home
  • 337099 - Ethyl 2-ethyl-2-hydroxy-3-oxobutyrate

EMAIL THIS PAGE TO A FRIEND
337099 Sigma-Aldrich

Ethyl 2-ethyl-2-hydroxy-3-oxobutyrate

96%

  • CAS Number 90113-78-3

  • Empirical Formula (Hill Notation) C8H14O4

  • Molecular Weight 174.19

  •  MDL number MFCD00075086

Purchase

Properties

assay   96%
Safety & Documentation

Safety Information

RIDADR 
UN 1993 / PGIII
WGK Germany 
3
Flash Point(F) 
188.6 °F
Flash Point(C) 
87 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?