• USA Home
  • 338907 - Iodo(trimethylphosphite)copper(I)

EMAIL THIS PAGE TO A FRIEND
338907 Sigma-Aldrich

Iodo(trimethylphosphite)copper(I)

98%

Synonym: Copper(I) iodide trimethyl phosphite complex, Cuprous iodide trimethylphosphite complex

Purchase

Properties

assay   98%
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?