• USA Home
  • 339857 - 4-(Trifluoromethoxy)benzonitrile

EMAIL THIS PAGE TO A FRIEND
339857 Sigma-Aldrich

4-(Trifluoromethoxy)benzonitrile

98%

Purchase

Properties

Related Categories Building Blocks, C8 to C9, Chemical Synthesis, Cyanides/Nitriles, Nitrogen Compounds - Organic Building Blocks,
Quality Level   200
assay   98%
refractive index   n20/D 1.452 (lit.)
bp   192-193 °C (lit.)
density   1.285 g/mL at 25 °C (lit.)
SMILES string   FC(F)(F)Oc1ccc(cc1)C#N
InChI   1S/C8H4F3NO/c9-8(10,11)13-7-3-1-6(5-12)2-4-7/h1-4H
InChI key   XWHIXOMWXCHJPP-UHFFFAOYSA-N

Description

General description

4-(Trifluoromethoxy)benzonitrile serves as an intermediate in the synthesis of fluvoxamine.

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Target organs 
Respiratory system
WGK Germany 
WGK 3
Flash Point(F) 
181.4 °F - closed cup
Flash Point(C) 
83 °C - closed cup

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?