• USA Home
  • 340278 - (S)-(+)-3-Hydroxy-4,4,4-trichlorobutyric β-lactone

EMAIL THIS PAGE TO A FRIEND
340278 Sigma-Aldrich

(S)-(+)-3-Hydroxy-4,4,4-trichlorobutyric β-lactone

99%

Synonym: β-Trichloromethyl-β-propiolactone, 4-(Trichloromethyl)-2-oxetanone

  • CAS Number 16493-63-3

  • Empirical Formula (Hill Notation) C4H3Cl3O2

  • Molecular Weight 189.42

  •  MDL number MFCD00075184

Purchase

Properties

assay   99%
optical purity   ee: 98% (GLC)
SMILES string   ClC(Cl)(Cl)[C@@H]1CC(=O)O1
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?