• USA Home
  • 340693 - 2-Amino-5-nitrobenzoic acid

EMAIL THIS PAGE TO A FRIEND
340693 Sigma-Aldrich

2-Amino-5-nitrobenzoic acid

technical grade, 90%

Synonym: 5-Nitroanthranilic acid

  • CAS Number 616-79-5

  • Linear Formula O2NC6H3(NH2)CO2H

  • Molecular Weight 182.13

  •  Beilstein/REAXYS Number 646219

  •  EC Number 210-493-6

  •  MDL number MFCD00017039

  •  PubChem Substance ID 24860845

Purchase

Properties

grade   technical grade
assay   90%
application(s)   peptide synthesis: suitable
mp   270 °C (dec.) (lit.)
SMILES string   Nc1ccc(cc1C(O)=O)[N+]([O-])=O
InChI   1S/C7H6N2O4/c8-6-2-1-4(9(12)13)3-5(6)7(10)11/h1-3H,8H2,(H,10,11)
InChI key   RUCHWTKMOWXHLU-UHFFFAOYSA-N

Description

Packaging

25 g in poly bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?