• USA Home
  • 340766 - Potassium phosphate tribasic

EMAIL THIS PAGE TO A FRIEND
340766 Aldrich

Potassium phosphate tribasic

97%

Synonym: Tripotassium phosphate

Purchase

Properties

Related Categories Essential Chemicals, Potassium Salts, Research Essentials, Salts
InChI Key   LWIHDJKSTIGBAC-UHFFFAOYSA-K
assay   97%
density   2.564 g/mL at 25 °C(lit.)

Description

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
TC8450000

Same Products - New Packaging
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P5629 Potassium phosphate tribasic, reagent grade, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?