EMAIL THIS PAGE TO A FRIEND
341797 Sigma-Aldrich

1-Phenyl-1-butyne

99%

Synonym: 1-Butynylbenzene

Purchase

Properties

Related Categories Alkynes, Building Blocks, Chemical Synthesis, Internal, Organic Building Blocks More...
InChI Key   FFFMSANAQQVUJA-UHFFFAOYSA-N
assay   99%
refractive index   n20/D 1.551 (lit.)
bp   73-75 °C/4 mmHg(lit.)
density   0.916 g/mL at 25 °C (lit.)

Description

General description

1-Phenyl-1-butyne is a phenyl alkyl acetylenic compound. It′s reaction with alkali metals (sodium, potassium) in various solvents is reported. Palladium particles incorporated into organophilic montmorillonite catalyzed liquid-phase hydrogenation of 1-phenyl-1-butyne is reported. Copolymerization of C60 and 1-phenyl-1-butyne is reported.

Application

1-Phenyl-1-butyne was used to investigate the liquid-phase stereoselective hydrogenation of phenyl alkyl acetylenics at 298K and atmospheric pressure on Pd-supported catalysts.

Packaging

5, 25 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
156.2 °F
Flash Point(C) 
69 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles

Related Content

Alkynes and Acetylenic Building Blocks

We list over 600 alkyne building blocks, including a wide array of propargyl alcohols, acetylenic boron reagents, and halogenated substrates for cross-coupling. Additionally, many of these are termin...
Keywords: Building blocks, Coupling reactions, Cross couplings

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?