• USA Home
  • 342351 - Hydroxylamine phosphate salt (3:1)

EMAIL THIS PAGE TO A FRIEND
342351 Sigma-Aldrich

Hydroxylamine phosphate salt (3:1)

97%

Purchase

Properties

Related Categories C-X Bond Formation (Non-Halogen), Chemical Synthesis, Other Non-Halogen Reagents for C-X Bond Formation, Synthetic Reagents
assay   97%
mp   169-171 °C (lit.)
storage temp.   room temp
SMILES string   NO.NO.NO.OP(O)(O)=O
InChI   1S/3H3NO.H3O4P/c3*1-2;1-5(2,3)4/h3*2H,1H2;(H3,1,2,3,4)
InChI key   XBUFCZMOAHHGMX-UHFFFAOYSA-N

Description

Application

Reactant for:
• Aqueous preparation of rhenium diphosphate
• Gas and solid state reactions with acetone
• Hydroxylamine formation via photolysis

Biochem/physiol Actions

MAO inhibitor; inhibits platelet aggregation

Safety & Documentation

Safety Information

Signal word 
Danger
RIDADR 
UN3262 - class 8 - PG 2 - EHS - Corrosive solid, basic, inorganic, n.o.s., HI: all
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?