• USA Home
  • 343498 - trans-2,4-Pentadienoic acid

EMAIL THIS PAGE TO A FRIEND
343498 Aldrich

trans-2,4-Pentadienoic acid

technical grade, 80%

  • CAS Number 21651-12-7

  • Empirical Formula (Hill Notation) C5H6O2

  • Molecular Weight 98.10

  •  MDL number MFCD00014017

Purchase

Properties

grade   technical grade
assay   80%
impurities   10% polymeric material
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?