• USA Home
  • 344222 - (1S)-(−)-Camphor p-tosylhydrazone

EMAIL THIS PAGE TO A FRIEND
344222 Sigma-Aldrich

(1S)-(−)-Camphor p-tosylhydrazone

  • CAS Number 123408-99-1

  • Empirical Formula (Hill Notation) C17H24N2O2S

  • Molecular Weight 320.45

  •  MDL number MFCD00075291

Purchase

Properties

optical activity   [α]20/D −8.5°, c = 1 in acetone
mp   164-167 °C (lit.)
SMILES string   Cc1ccc(cc1)S(=O)(=O)N\N=C2/C[C@@H]3CC[C@@]2(C)C3(C)C
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?