• USA Home
  • 344338 - Bis(trimethylsilyl)carbodiimide

EMAIL THIS PAGE TO A FRIEND
344338 Sigma-Aldrich

Bis(trimethylsilyl)carbodiimide

98%

Synonym: 1,3-Bis(trimethylsilyl)carbodiimide, N,N′-Methanetetraylbis[1,1,1-trimethylsilylamine], N,N-Bis(trimethylsilyl)carbodiimide

  • CAS Number 1000-70-0

  • Linear Formula (CH3)3SiN=C=NSi(CH3)3

  • Molecular Weight 186.40

  •  Beilstein/REAXYS Number 2042082

  •  EC Number 213-673-2

  •  MDL number MFCD00051538

  •  PubChem Substance ID 24861243

  •  NACRES NA.22

Purchase

Properties

Related Categories C-X Bond Formation (Non-Halogen), Chemical Synthesis, Other Non-Halogen Reagents for C-X Bond Formation, Synthetic Reagents
Quality Level   200
assay   98%
refractive index   n20/D 1.434 (lit.)
bp   164 °C (lit.)
mp   −23 °C (lit.)
density   0.821 g/mL at 25 °C (lit.)
SMILES string   C[Si](C)(C)N=C=N[Si](C)(C)C
InChI   1S/C7H18N2Si2/c1-10(2,3)8-7-9-11(4,5)6/h1-6H3
InChI key   KSVMTHKYDGMXFJ-UHFFFAOYSA-N

Description

Application

Reactant for preparation of:
• 4-substituted methoxylbenzoyl-aryl-thiazole analogues as potent and orally bioavailable anticancer agents
• O-silylurethanes
• Porous SiCN materials
• Luminescent diketo-pyrrolo-pyrrole analog
• Hard thin films of silicon carbonitride SiCN as potential protection coatings against wear and corrosion of metals
• Ceramic materials

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Target organs 
Respiratory system
RIDADR 
UN 1993BE 3 / PGII
WGK Germany 
WGK 3
RTECS 
VV1302500
Flash Point(F) 
69.8 °F - closed cup
Flash Point(C) 
21 °C - closed cup
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?