EMAIL THIS PAGE TO A FRIEND
344362 Aldrich

Diethyl ether

ACS reagent, anhydrous, ≥99%

Synonym: Ether, Ethyl ether

  • CAS Number 60-29-7

  • Linear Formula (CH3CH2)2O

  • Molecular Weight 74.12

  •  Beilstein/REAXYS Number 1696894

  •  MDL number MFCD00011646

Purchase

Properties

grade   ACS reagent
  anhydrous
vapor density   2.6 (vs air)
InChI Key   RTZKZFJDLAIYFH-UHFFFAOYSA-N
assay   ≥99%
autoignition temp.   320 °F
contains   1 ppm BHT as stabilizer
expl. lim.   36.5 %
impurities   ≤0.001% carbonyl
  ≤0.03% water
  0.0002 meq/g Titr. Acid
evapn. residue   ≤0.001%
color   APHA: ≤10
refractive index   n20/D 1.3530 (lit.)
bp   34.6 °C(lit.)
mp   −116 °C (lit.)
density   0.706 g/mL at 25 °C (lit.)

Description

Packaging

Packaged in safety cans

Other Notes

Contains peroxide

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Pressure-Temperature Calculator for Solvents

*Calculations provide estimated results based on Antoine's Coefficients and their associated temperature range values. Antoine's Coefficients were obtained from the National Institute of Standards an...

Related Content

Pressure-Temperature Nomograph Interactive Tool

Use the interactive controls above to simplify calculations and improve the efficiency of your distillation or evaporation requirements.
Keywords: Evaporation, Vaporization

Properties of Solvents Table

The table below presents the physical characteristics of common solvents. Easily reference or compare solvent properties like boiling point, density, and refractive index all from one list.
Keywords: Gas chromatography, Green chemistry, High performance liquid chromatography, Melting, Solvents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?