• USA Home
  • 344753 - 1-Piperidinecarboxamide

EMAIL THIS PAGE TO A FRIEND
344753 Sigma-Aldrich

1-Piperidinecarboxamide

99%

Synonym: 1-Carbamylpiperidine

Purchase

Properties

Related Categories Building Blocks, C5 to C7, Chemical Synthesis, Heterocyclic Building Blocks, Piperidines More...
assay   99%
mp   104-106 °C (lit.)
SMILES string   NC(=O)N1CCCCC1
InChI   1S/C6H12N2O/c7-6(9)8-4-2-1-3-5-8/h1-5H2,(H2,7,9)
InChI key   JSPCTNUQYWIIOT-UHFFFAOYSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
TM5975700

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?