• USA Home
  • 346535 - 2,2-Dimethyl-5-(1H-pyrrol-2-ylmethylene)-1,3-dioxane-4,6-dione

EMAIL THIS PAGE TO A FRIEND
346535 Aldrich

2,2-Dimethyl-5-(1H-pyrrol-2-ylmethylene)-1,3-dioxane-4,6-dione

99%

  • CAS Number 23111-03-7

  • Empirical Formula (Hill Notation) C11H11NO4

  • Molecular Weight 221.21

  •  MDL number MFCD00075361

Purchase

Properties

assay   99%
mp   180-182 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?