• USA Home
  • 346659 - 4-Nitropyridine N-oxide

EMAIL THIS PAGE TO A FRIEND
346659 Sigma-Aldrich

4-Nitropyridine N-oxide

97%

Purchase

Properties

Related Categories Building Blocks, C5, Chemical Synthesis, Heterocyclic Building Blocks, Pyridines More...
InChI Key   RXKNNAKAVAHBNK-UHFFFAOYSA-N
assay   97%
mp   159-162 °C (lit.)

Description

General description

4-Nitropyridine N-oxide has been reported as a quorum-sensing inhibitor, having IC50 value of 24μg/ml. Facile one-pot arylation and alkylation of 4-nitropyridine N-oxide has been reported. 4-Nitropyridine N-oxide can be prepared from pyridine by oxidation with peracetic acid and nitration. Primary photochemical process of 4-nitropyridine N-oxide has been investigated by steady-light irradiation and a flash spectroscopic method.

Application

4-Nitropyridine N-oxide may be used in the preparation of several 4-substituted pyridine-N-oxides and pyridines.

Packaging

25, 100 g in poly bottle

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
UT6380000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?