• USA Home
  • 346918 - Poly(ethylene-co-vinyl acetate)

EMAIL THIS PAGE TO A FRIEND
346918 Sigma-Aldrich

Poly(ethylene-co-vinyl acetate)

vinyl acetate 33 wt. %, melt index 43 g/10 min (190°C/2.16 kg)

Purchase

Properties

form   pellets
autoignition temp.   500 °F
melt index   43 g/10 min (190°C/2.16 kg)
composition   vinyl acetate, 33 wt. %
SMILES string   C=C.CC(=O)OC=C
InChI   1S/C4H6O2.C2H4/c1-2-3-4(5)6;1-2/h2H,1,3H2,(H,5,6);1-2H2
InChI key   DQXBYHZEEUGOBF-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?