EMAIL THIS PAGE TO A FRIEND
347140 Aldrich

Titanium

granular, −15 mesh, 99.95%

Purchase

Properties

assay   99.95%
form   granular
particle size   −15 mesh
Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2546 4.2 / PGII
WGK Germany 
nwg
RTECS 
XR1700000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?