EMAIL THIS PAGE TO A FRIEND
34810 Aldrich

Dibutyl phosphate

purum, ≥97.0% (T)

Synonym: Phosphoric acid dibutyl ester

  • CAS Number 107-66-4

  • Linear Formula (CH3CH2CH2CH2O)2P(O)OH

  • Molecular Weight 210.21

  •  Beilstein/REAXYS Number 607224

  •  EC Number 203-509-8

  •  MDL number MFCD00015245

  •  PubChem Substance ID 24861550

Purchase

Properties

InChI Key   JYFHYPJRHGVZDY-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (T)
refractive index   n20/D 1.428
density   1.06 g/mL at 20 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3265 8 / PGII
WGK Germany 
1
RTECS 
TB9605000
Flash Point(F) 
314.6 °F
Flash Point(C) 
157 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

68572 Dibutyl phosphate, ≥97.0% (T)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?