• USA Home
  • 348201 - 1-Bromo-4-(trimethylsiloxy)benzene

EMAIL THIS PAGE TO A FRIEND
348201 Sigma-Aldrich

1-Bromo-4-(trimethylsiloxy)benzene

98%

Synonym: (4-Bromophenoxy)trimethylsilane

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds, Protected Alcohols/Phenols More...
assay   98%
refractive index   n20/D 1.515 (lit.)
density   1.268 g/mL at 25 °C (lit.)
SMILES string   C[Si](C)(C)Oc1ccc(Br)cc1
InChI   1S/C9H13BrOSi/c1-12(2,3)11-9-6-4-8(10)5-7-9/h4-7H,1-3H3
InChI key   HWKPTBHJWBSOOP-UHFFFAOYSA-N

Description

General description

1-Bromo-4-(trimethylsiloxy)benzene is a protected phenol.

Application

1-Bromo-4-(trimethylsiloxy)benzene (4-Bromophenoxy)trimethylsilane) may be used in the synthesis of 4-bromophenyl-β-O-D-tetraacetylglucoside.

Packaging

25 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
206.6 °F
Flash Point(C) 
97 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?