EMAIL THIS PAGE TO A FRIEND
348570 Sigma-Aldrich

Cadmium

foil, thickness 0.25 mm, ≥99.99%

  • CAS Number 7440-43-9

  • Empirical Formula (Hill Notation) Cd

  • Molecular Weight 112.41

  •  Beilstein/REAXYS Number 8137359

  •  EC Number 231-152-8

  •  MDL number MFCD00010914

Purchase

Properties

assay   ≥99.99%
form   foil
resistivity   7.27 μΩ-cm, 22°C
thickness   0.25 mm
bp   765 °C (lit.)
mp   320.9 °C (lit.)
density   8.65 g/mL at 25 °C (lit.)
SMILES string   [Cd]
InChI   1S/Cd
InChI key   BDOSMKKIYDKNTQ-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?