EMAIL THIS PAGE TO A FRIEND
348740 Aldrich

Silver

foil, thickness 0.05 mm, 99.9% trace metals basis

Purchase

Properties

InChI Key   BQCADISMDOOEFD-UHFFFAOYSA-N
assay   99.9% trace metals basis
form   foil
resistivity   1.59 μΩ-cm, 20°C
thickness   0.05 mm
bp   2212 °C(lit.)
mp   960 °C (lit.)
density   10.49 g/cm3(lit.)

Description

Quantity

1.3 g = 50 × 50 mm; 5.2 g = 100 × 100 mm

Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Signal word 
Warning
Hazard statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?