• USA Home
  • 348880 - 1,2-Dibromo-3,3-dimethylbutane

EMAIL THIS PAGE TO A FRIEND
348880 Sigma-Aldrich

1,2-Dibromo-3,3-dimethylbutane

97%

Purchase

Properties

assay   97%
refractive index   n20/D 1.5053 (lit.)
bp   73 °C/3 mmHg (lit.)
density   1.61 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993 / PGIII
WGK Germany 
3
Flash Point(F) 
183.2 °F
Flash Point(C) 
84 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?