EMAIL THIS PAGE TO A FRIEND
349518 Sigma-Aldrich

4-Bromo-2,6-xylenol

≥99.5%

Synonym: 4-Bromo-2,6-dimethylphenol

Purchase

Properties

InChI Key   ZLVFYUORUHNMBO-UHFFFAOYSA-N
assay   ≥99.5%
mp   74-78 °C (lit.)
Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
ZE6780000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

196371 4-Bromo-2,6-xylenol, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?