EMAIL THIS PAGE TO A FRIEND
356921 Sigma-Aldrich

Lead

foil, thickness 0.127 mm, ≥99.99%

Synonym: Lead element

  • CAS Number 7439-92-1

  • Empirical Formula (Hill Notation) Pb

  • Molecular Weight 207.20

  •  MDL number MFCD00134050

Purchase

Properties

assay   ≥99.99%
form   foil
resistivity   20.65 μΩ-cm
thickness   0.127 mm
bp   1740 °C (lit.)
mp   327.4 °C (lit.)
SMILES string   [Pb]
InChI   1S/Pb
InChI key   WABPQHHGFIMREM-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?