EMAIL THIS PAGE TO A FRIEND
356980 Sigma-Aldrich

Tantalum

rod, diam. 3.2 mm, ≥99.9% trace metals basis

Synonym: Ta

Purchase

Properties

Related Categories Materials Science, Metal and Ceramic Science, Metals, Tantalum
vapor pressure   <0.01 mmHg ( 537.2 °C)
InChI Key   GUVRBAGPIYLISA-UHFFFAOYSA-N
assay   ≥99.9% trace metals basis
form   rod
autoignition temp.   572 °F
resistivity   13.5 μΩ-cm, 20°C
diam.   3.2 mm
bp   5425 °C(lit.)
mp   2996 °C (lit.)
density   16.69 g/cm3(lit.)

Description

Packaging

6.5 g in poly tube

Quantity

6.5 g = 5 cm; 32.5 g = 25 cm

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
Protocols & Articles

Articles

Biomedical Implant Devices Fabricated from Low Young’s Modulus Titanium Alloys Demonstrating High Mechanical Biocompatibility

Mitsuo Niinomi, Masasaki Nakai* Institute for Materials Research, Tohoku University 2-1-1, Katahira, Aoba-ku, Sendai 8577, Japan *not pictured Email: niinomi@imr.tohoku.ac.jp

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?