EMAIL THIS PAGE TO A FRIEND
357014 Sigma-Aldrich

Lead

wire, diam. 0.5 mm, 99.999% trace metals basis

Synonym: Lead element

Purchase

Properties

assay   99.999% trace metals basis
form   wire
resistivity   20.65 μΩ-cm
diam.   0.5 mm
bp   1740 °C (lit.)
mp   327.4 °C (lit.)
SMILES string   [Pb]
InChI   1S/Pb
InChI key   WABPQHHGFIMREM-UHFFFAOYSA-N

Description

Quantity

2.2 g = 1 m; 11 g = 5 m

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
nwg
RTECS 
OF7525000
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?