• USA Home
  • 359203 - Tetrapentylammonium iodide

EMAIL THIS PAGE TO A FRIEND
359203 Aldrich

Tetrapentylammonium iodide

98%

  • CAS Number 2498-20-6

  • Linear Formula [CH3(CH2)4]4N(I)

  • Molecular Weight 425.47

  •  Beilstein Registry Number 3917412

  •  MDL number MFCD00041980

Purchase

Properties

assay   98%
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?