EMAIL THIS PAGE TO A FRIEND
359696 Sigma-Aldrich

Boc-Ser-OH

98%

Synonym: N-(tert-Butoxycarbonyl)-L-serine, Boc-L-serine

  • CAS Number 3262-72-4

  • Linear Formula HOCH2CH(COOH)NHCOOC(CH3)3

  • Molecular Weight 205.21

  •  Beilstein/REAXYS Number 2212252

  •  EC Number 221-867-3

  •  MDL number MFCD00037243

  •  PubChem Substance ID 24862099

Purchase

Properties

assay   98%
optical activity   [α]20/D −3°, c = 2 in acetic acid
mp   91 °C (dec.) (lit.)
SMILES string   CC(C)(C)OC(=O)N[C@@H](CO)C(O)=O
InChI   1S/C8H15NO5/c1-8(2,3)14-7(13)9-5(4-10)6(11)12/h5,10H,4H2,1-3H3,(H,9,13)(H,11,12)/t5-/m0/s1
InChI key   FHOAKXBXYSJBGX-YFKPBYRVSA-N

Description

Application

Building block in peptide synthesis. Starting material for the synthesis of various α-amino acids via the β-lactone.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

15500 Boc-Ser-OH, ≥99.0% (T)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?