• USA Home
  • 360201 - L-Leucine-4-nitroanilide

EMAIL THIS PAGE TO A FRIEND
360201 Sigma-Aldrich

L-Leucine-4-nitroanilide

99%

Synonym: L-Leucine p-nitroanilide Free Base, L-Leucine p-nitroanilide

  • CAS Number 4178-93-2

  • Linear Formula (CH3)2CHCH2CH(NH2)CONHC6H4NO2

  • Molecular Weight 251.28

  •  Beilstein/REAXYS Number 2218433

  •  EC Number 224-047-3

  •  MDL number MFCD00063681

Purchase

Properties

assay   99%
optical activity   [α]20/D +96°, c = 1 in 1 M HCl
mp   88-90 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

L9125 L-Leucine-p-nitroanilide, leucine aminopeptidase substrate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?