EMAIL THIS PAGE TO A FRIEND
360422 Aldrich

α-Ecdysone

97%

Synonym: 2β,3β,14α,22(R),25-Pentahydroxy-7-cholesten-6-one

  • CAS Number 3604-87-3

  • Empirical Formula (Hill Notation) C27H44O6

  • Molecular Weight 464.63

  •  Beilstein/REAXYS Number 2422986

  •  MDL number MFCD00042683

Purchase

Properties

InChI Key   UPEZCKBFRMILAV-JMZLNJERSA-N
assay   97%
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?