• USA Home
  • 360678 - 2-(3-Nitrophenylsulfonyl)ethanol

EMAIL THIS PAGE TO A FRIEND
360678 Aldrich

2-(3-Nitrophenylsulfonyl)ethanol

97%

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, N, Stains & Dyes, A to Z, Stains and Dyes More...
InChI Key   VMORQDKKMBAQPJ-UHFFFAOYSA-N
assay   97%
mp   76-78 °C(lit.)

Description

Packaging

1, 5 g in glass bottle

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?