• USA Home
  • 360813 - Ethyl (Z)-(hydroxyimino)[2-(tritylamino)thiazol-4-yl]acetate hydrochloride

EMAIL THIS PAGE TO A FRIEND
360813 Sigma-Aldrich

Ethyl (Z)-(hydroxyimino)[2-(tritylamino)thiazol-4-yl]acetate hydrochloride

97%

Synonym: Ethyl (Z)-[2-(tritylamino)thiazol-4-yl](hydroxyimino)acetate hydrochloride

  • CAS Number 66339-00-2

  • Empirical Formula (Hill Notation) C26H23N3O3S · HCl

  • Molecular Weight 494.01

  •  Beilstein/REAXYS Number 5697264

  •  EC Number 266-321-5

  •  MDL number MFCD00062755

Purchase

Properties

assay   97%
mp   176-180 °C (dec.) (lit.)
SMILES string   Cl.CCOC(=O)C(=N/O)\c1csc(NC(c2ccccc2)(c3ccccc3)c4ccccc4)n1
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?