• USA Home
  • 360856 - S-2-Benzothiazolyl-2-amino-α-(methoxyimino)-4-thiazolethiolacetate

EMAIL THIS PAGE TO A FRIEND
360856 Sigma-Aldrich

S-2-Benzothiazolyl-2-amino-α-(methoxyimino)-4-thiazolethiolacetate

97%

Purchase

Properties

assay   97%
mp   128-130 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
XJ4664500
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?