• USA Home
  • 361453 - 3,3,5,5-Tetramethyl-1,2-cyclopentanedione

EMAIL THIS PAGE TO A FRIEND
361453 Aldrich

3,3,5,5-Tetramethyl-1,2-cyclopentanedione

99%

  • CAS Number 20633-06-1

  • Empirical Formula (Hill Notation) C9H14O2

  • Molecular Weight 154.21

  •  MDL number MFCD00134193

Purchase

Properties

assay   99%
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?