• USA Home
  • 361542 - 4-Amino-4-(3-hydroxypropyl)-1,7-heptanediol

EMAIL THIS PAGE TO A FRIEND
361542 Sigma-Aldrich

4-Amino-4-(3-hydroxypropyl)-1,7-heptanediol

97%

Synonym: Bis-homotris

Purchase

Properties

assay   97%
bp   220-235 °C/0.5 mmHg (lit.)
mp   108-110 °C (lit.)
SMILES string   NC(CCCO)(CCCO)CCCO
InChI   1S/C10H23NO3/c11-10(4-1-7-12,5-2-8-13)6-3-9-14/h12-14H,1-9,11H2
InChI key   HACQKMWYOKGLET-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?