• USA Home
  • 361917 - trans-4-Hydroxy-3-methoxy-β-nitrostyrene

EMAIL THIS PAGE TO A FRIEND
361917 Sigma-Aldrich

trans-4-Hydroxy-3-methoxy-β-nitrostyrene

98%

Synonym: trans-2-Methoxy-4-(2-nitrovinyl)phenol

Purchase

Properties

Related Categories Acyclic, Alkenes, Building Blocks, Chemical Synthesis, Organic Building Blocks More...
InChI Key   XVXCSPJSXIUNQJ-SNAWJCMRSA-N
assay   98%
mp   168-170 °C (lit.)
Gene Information   human ... EGFR(1956)

Description

General description

trans-4-Hydroxy-3-methoxy-β-nitrostyrene is a phenolic β-nitrostyrene. Synthesis of 4-hydroxy-3-methoxy-β-nitrostyrene is reported.

Application

trans-4-Hydroxy-3-methoxy-β-nitrostyrene may be used in the synthesis of cytotoxic β-nitrostyrene derivatives, 4-O-benzoyl-3-methoxy-β-nitrostyrene (BMNS) derivatives.

Packaging

25 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?