• USA Home
  • 361992 - (−)-Caryophyllene oxide

EMAIL THIS PAGE TO A FRIEND
361992 Sigma-Aldrich

(−)-Caryophyllene oxide

technical grade, 90%

Synonym: β-Caryophyllene epoxide, (−)-Epoxycaryophyllene, (−)-Epoxydihydrocaryophyllene, (1R,4R,6R,10S)-9-Methylene-4,12,12-trimethyl-5-oxatricyclo[8.2.0.04,6]dodecane, trans-Caryophyllene oxide

  • CAS Number 1139-30-6

  • Empirical Formula (Hill Notation) C15H24O

  • Molecular Weight 220.35

  •  Beilstein/REAXYS Number 148213

  •  EC Number 214-519-7

  •  MDL number MFCD00134216

  •  PubChem Substance ID 24862287

Purchase

Properties

grade   technical grade
InChI Key   NVEQFIOZRFFVFW-RGCMKSIDSA-N
assay   90%
optical activity   [α]20/D −57°, c = 2 in chloroform
mp   62-63 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
RP5530000
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

22076 (−)-Caryophyllene oxide, ≥99.0% (sum of enantiomers, GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?