• USA Home
  • 362298 - Trimethyl cis,cis-1,3,5-cyclohexanetricarboxylate

EMAIL THIS PAGE TO A FRIEND
362298 Sigma-Aldrich

Trimethyl cis,cis-1,3,5-cyclohexanetricarboxylate

98%

Purchase

Properties

Related Categories Building Blocks, C12 to C63, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   ZZNOMLSSCRBRJS-AYMMMOKOSA-N
assay   98%
mp   46-49 °C (lit.)

Description

Packaging

5 g in glass bottle

Application

Trimethyl cis,cis-1,3,5-cyclohexanetricarboxylate is suitable for use in the preparation of tripropyl imide acid. It is also suitable for use in the preparation of lipophilic analog of Kemp′s acid.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C

Documents

Certificate of Analysis

Certificate of Origin


Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?