• USA Home
  • 362786 - (S)-(−)-2-Acetoxypropionic acid

EMAIL THIS PAGE TO A FRIEND
362786 Aldrich

(S)-(−)-2-Acetoxypropionic acid

98%

Synonym: O-Acetyl-L-lactic acid

  • CAS Number 6034-46-4

  • Linear Formula CH3CH(O2CCH3)CO2H

  • Molecular Weight 132.11

  •  MDL number MFCD00191370

Purchase

Properties

assay   98%
optical activity   [α]20/D −51°, c = 7 in chloroform
refractive index   n20/D 1.422 (lit.)
bp   115-117 °C/2 mmHg(lit.)
density   1.162 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
235 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?