• USA Home
  • 362921 - Diethyl 2,2′-(1,3-dithietane-2,4-diylidene)bis(cyanoacetate)

EMAIL THIS PAGE TO A FRIEND
362921 Sigma-Aldrich

Diethyl 2,2′-(1,3-dithietane-2,4-diylidene)bis(cyanoacetate)

98%

  • CAS Number 22624-54-0

  • Empirical Formula (Hill Notation) C12H10N2O4S2

  • Molecular Weight 310.35

  •  MDL number MFCD00205376

  •  PubChem Substance ID 24862340

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Other Heterocyclic Building Blocks, S-Containing More...
InChI Key   SLORFIRBIOJAPR-NTLHZVPKSA-N
assay   98%
mp   165-168 °C (lit.)

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?