• USA Home
  • 363618 - 1,5-Dihydro-3-methoxy-2,4-benzodioxepin

EMAIL THIS PAGE TO A FRIEND
363618 Sigma-Aldrich

1,5-Dihydro-3-methoxy-2,4-benzodioxepin

98%

  • CAS Number 67461-24-9

  • Empirical Formula (Hill Notation) C10H12O3

  • Molecular Weight 180.20

  •  MDL number MFCD00134264

Purchase

Properties

assay   98%
bp   75-78 °C/0.2 mmHg (lit.)
mp   42-44 °C (lit.)

Description

Application

Protecting group for carbonyl compounds that can be removed under nonacidic conditions (i.e., by catalytic hydrogenolysis).

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?