• USA Home
  • 363863 - 2,3,5-Tri-O-benzyl-1-O-(4-nitrobenzoyl)-D-arabinofuranose, predominantly α anomer

EMAIL THIS PAGE TO A FRIEND
363863 Sigma-Aldrich

2,3,5-Tri-O-benzyl-1-O-(4-nitrobenzoyl)-D-arabinofuranose, predominantly α anomer

97%

Purchase

Properties

assay   97%
optical activity   [α]20/D +57°, c = 1.2 in methylene chloride
mp   86-88 °C (lit.)
SMILES string   [O-][N+](=O)c1ccc(cc1)C(=O)OC2O[C@H](OCc3ccccc3)[C@@H](OCc4ccccc4)[C@@H]2OCc5ccccc5
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?