EMAIL THIS PAGE TO A FRIEND
364118 Sigma-Aldrich

1-Aza-18-crown-6

95%

Synonym: 1,4,7,10,13-Pentaoxa-16-azacyclooctadecane

  • CAS Number 33941-15-0

  • Empirical Formula (Hill Notation) C12H25NO5

  • Molecular Weight 263.33

  •  Beilstein/REAXYS Number 1074079

  •  EC Number 251-751-8

  •  MDL number MFCD00075466

  •  PubChem Substance ID 24862414

Purchase

Properties

assay   95%
mp   46-49 °C (lit.)
SMILES string   C1COCCOCCOCCOCCOCCN1
InChI   1S/C12H25NO5/c1-3-14-5-7-16-9-11-18-12-10-17-8-6-15-4-2-13-1/h13H,1-12H2
InChI key   NBXKUSNBCPPKRA-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

11382 1-Aza-18-crown-6, ≥98.0% (NT)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?