EMAIL THIS PAGE TO A FRIEND
364800 Sigma-Aldrich

Disperse Orange 13

Dye content 90 %

Synonym: 4-[4-(Phenylazo)-1-naphthylazo]phenol

  • CAS Number 6253-10-7

  • Linear Formula C6H5N=NC10H6N=NC6H4OH

  • Molecular Weight 352.39

  •  Colour Index Number 26080

  •  EC Number 228-381-0

  •  MDL number MFCD00003909

  •  PubChem Substance ID 24862456

Purchase

Properties

Related Categories Cell Biology, D, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes More...
composition   Dye content, 90%
mp   149-153 °C (lit.)
λmax   427 nm
SMILES string   Oc1ccc(cc1)\N=N\c2ccc(\N=N\c3ccccc3)c4ccccc24
InChI   1S/C22H16N4O/c27-18-12-10-17(11-13-18)24-26-22-15-14-21(19-8-4-5-9-20(19)22)25-23-16-6-2-1-3-7-16/h1-15,27H/b25-23+,26-24+
InChI key   FWTBRYBHCBCJEQ-OGGGYYITSA-N

Description

Packaging

1, 5 g in glass bottle

Features and Benefits

Nonlinear optics

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?