• USA Home
  • 364894 - (S)-(+)-3-Benzyloxy-1,2-propanediol 1-(p-toluenesulfonate)

EMAIL THIS PAGE TO A FRIEND
364894 Aldrich

(S)-(+)-3-Benzyloxy-1,2-propanediol 1-(p-toluenesulfonate)

95%

Synonym: (S)-(+)-1-Benzyloxy-3-tosyloxy-2-propanol, 3-Benzyl-sn-glycerol 1-tosylat

  • CAS Number 16495-04-8

  • Empirical Formula (Hill Notation) C17H20O5S

  • Molecular Weight 336.40

Purchase

Properties

assay   95%
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?