• USA Home
  • 365262 - Ethyl dimethylphosphonoacetate

EMAIL THIS PAGE TO A FRIEND
365262 Sigma-Aldrich

Ethyl dimethylphosphonoacetate

98%

Synonym: Ethoxycarbonylmethylphosphonic acid dimethylester, Phosphonoacetic acid P,P-dimethyl ethylester

  • CAS Number 311-46-6

  • Linear Formula (CH3O)2P(O)CH2COOC2H5

  • Molecular Weight 196.14

  •  Beilstein/REAXYS Number 1777603

  •  EC Number 206-222-6

  •  MDL number MFCD00015677

Purchase

Properties

assay   98%
refractive index   n20/D 1.435 (lit.)
bp   134 °C/10 mmHg (lit.)
density   1.188 g/mL at 25 °C (lit.)
SMILES string   CCOC(=O)CP(=O)(OC)OC
InChI   1S/C6H13O5P/c1-4-11-6(7)5-12(8,9-2)10-3/h4-5H2,1-3H3
InChI key   HUNISAHOCCASGM-UHFFFAOYSA-N

Description

Application

Reactant for synthesis of:
• Non-nucleoside inhibitors of HCV polymerase NS5B
• Neuropeptide Y1 receptor antagonists
• Alkenylcarboxylates from Horner-Wadsworth-Emmons reactions

Reactant for:
• Enantioselective transfer hydrogenation for preparation of the C7-C14 fragment of ulapualide A
• Intramolecular Michael reactions
• Olefination of peptidyl aldehydes with stabilized ylides

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
210.2 °F
Flash Point(C) 
99 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

02610 Ethyl dimethylphosphonoacetate, technical, ≥90% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?