• USA Home
  • 366323 - 4,4′-Diphenyl-2,2′-dipyridyl

EMAIL THIS PAGE TO A FRIEND
366323 Sigma-Aldrich

4,4′-Diphenyl-2,2′-dipyridyl

technical grade

Purchase

Properties

grade   technical grade
mp   188-190 °C (lit.)
SMILES string   c1ccc(cc1)-c2ccnc(c2)-c3cc(ccn3)-c4ccccc4
InChI   1S/C22H16N2/c1-3-7-17(8-4-1)19-11-13-23-21(15-19)22-16-20(12-14-24-22)18-9-5-2-6-10-18/h1-16H
InChI key   OXMSMRJQZMTIMT-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?