• USA Home
  • 366749 - 4,6-Dinitropyrogallol

EMAIL THIS PAGE TO A FRIEND
366749 Sigma-Aldrich

4,6-Dinitropyrogallol

97%

Purchase

Properties

assay   97%
mp   209-211 °C (lit.)
SMILES string   Oc1c(O)c(cc(c1O)[N+]([O-])=O)[N+]([O-])=O
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?